missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isonicotinamide-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Isonicotinamide-2,3,5,6-d4 |
| CAS | 1219799-40-2 |
| Molecular Formula | C6 2H4 H2 N2 O |
| Quantity | 0.25 g |
| Synonym | Isonicotinamide-d4, Isonicotinamide-2,3,5,6-D4, 4-Pyridinecarboxamide-2,3,5,6-d4, 4-(Aminocarbonyl)-2,3,5,6-tetradeuteriopyridine |
| InChI Formula | InChI=1 S/C6H6N2O/c7-6(9)5-1-3-8-4-2-5/h1-4 H,(H2,7,9)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1nc([2 H])c([2 H])c(C(=O)N)c1[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuteriopyridine-4-carboxamide |
| Molecular Weight (g/mol) | 126.15 |
| Formula Weight | 126.0731 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction