missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine-d7 (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | L-Leucine-4,5,5,5,5',5',5'-D7 |
| CAS | 92751-17-2 |
| Molecular Formula | C6 D7 H6 N O2 |
| Quantity | 0.25 g |
| Synonym | L-Leucine-4,5,5,5,5',5',5'-d7 (9 CI, ACI), L-Norvaline-4,5,5,5-d4, 4-(methyl-d3)- (ZCI), L-Leucine-d7 (iso-propyl-d7), L-Leucine-d7, (S)-2-Amino-(4,5,5,5-tetradeuterio)-4-(trideuteriomethyl)pentanoic acid, L-Leucine-4,5,5,5,5',5',5'-D7, L-Leucine-4,5,5,5,5',5',5'-D7 |
| InChI Formula | InChI=1 S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i1D3,2D3,4 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C[C@H](N)C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | (2 S)-2-amino-4,5,5,5-tetradeuterio-4-(trideuteriomethyl)pentanoic acid |
| Molecular Weight (g/mol) | 138.2161 |
| Formula Weight | 138.1386 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction