missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine-d8-N-t-BOC, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | N-(tert-Butoxycarbonyl)-L-valine-2,3,4,4,4,4',4',4'-D8 |
| CAS | 153568-33-3 |
| Molecular Formula | C10 D8 H11 N O4 |
| Quantity | 0.25 g |
| Synonym | L-Valine-2,3,4,4,4,4',4',4'-d8, N-[(1,1-dimethylethoxy)carbonyl]- (9 CI), N-[(1,1-Dimethylethoxy)carbonyl]-L-valine-2,3,4,4,4,4',4',4'-d8, L-Valine-d8-N-t-BOC, N-[(1,1-Dimethylethoxy)carbonyl]-L-valine-d8, (2 S)-2-(tert-Butoxycarbonylamino)-3-(methyl-d3)butanoic-2,3,4,4,4-d5 acid, N-(tert-Butoxycarbonyl)-L-valine-2,3,4,4,4,4',4',4'-D8, N-(tert-Butoxycarbonyl)-L-valine-2,3,4,4,4,4',4',4'-D8 |
| InChI Formula | InChI=1 S/C10H19NO4/c1-6(2)7(8(12)13)11-9(14)15-10(3,4)5/h6-7 H,1-5H3,(H,11,14)(H,12,13)/t7-/m0/s1/i1D3,2D3,6 D,7 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])[C@]([2 H])(NC(=O)OC(C)(C)C)C(=O)O |
| IUPAC Name | (2 S)-2,3,4,4,4-pentadeuterio-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(trideuteriomethyl)butanoic acid |
| Molecular Weight (g/mol) | 225.3115 |
| Formula Weight | 225.1816 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction