missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Pentadecanoate-d29, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Pentadecanoic Acid Methyl Ester-D29 (pentadecanoic-D29) |
| CAS | 68060-02-6 |
| Molecular Formula | C16 D29 H3 O2 |
| Quantity | 0.25 g |
| Synonym | Pentadecanoic-d29 acid, methyl ester (9 CI), Pentadecanoic acid-d29 methyl ester, Methyl pentadecanoate-d29, Pentadecanoic Acid Methyl Ester-D29 (pentadecanoic-D29) |
| InChI Formula | InChI=1 S/C16H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18-2/h3-15H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC |
| IUPAC Name | methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,15-nonacosadeuteriopentadecanoate |
| Molecular Weight (g/mol) | 285.6028 |
| Formula Weight | 285.4223 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction