missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexadecylpyridinium-d5 Bromide, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Cetylpyridinium-D5 (pyridinium-D5) Bromide |
| CAS | 143715-91-7 |
| Molecular Formula | C21H33D5N . Br |
| Quantity | 0.25 g |
| Synonym | Pyridinium-2,3,4,5,6-d5, 1-hexadecyl-, bromide (9 CI), Cetylpyridinium-D5 (pyridinium-D5) bromide, 1-Hexadecyl-2,3,4,5,6-pentadeuteriopyridinium bromide, 1-Cetylpyridinium-d5 bromide;Cetylpyridinium-d5 bromide, Hexadecylpyridinium-d5 bromide, n-Hexadecylpy |
| InChI Formula | InChI=1 S/C21H38N.BrH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-22-20-17-15-18-21-22;/h15,17-18,20-21 H,2-14,16,19H2,1H3;1 H/q+1;/p-1/i15D,17 D,18 D,20 D,21 D; |
| SMILES | CCCCCCCCCCCCCCCC[N+](C([2 H])=C1[2 H])=C(C([2 H])=C1[2 H])[2 H].[Br-] |
| IUPAC Name | 1-hexadecylpyridin-1-ium-2,3,4,5,6-d5 bromide |
| Molecular Weight (g/mol) | 389.48 |
| Formula Weight | 388.25 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction