missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Tetradecyl-1,1-d2 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Tetradecanol-1,1-D2 |
| CAS | 169398-02-1 |
| Molecular Formula | C14 D2 H28 O |
| Quantity | 0.25 g |
| Synonym | 1-Tetradecan-1,1-d2-ol (9 CI), 1-Tetradecanol-1,1-d2, 1-Tetradecanol-1,1-D2, Myristyl alcohol-d2, n-Tetradecyl-1,1-d2 alcohol, Tetradecanol-d2 |
| InChI Formula | InChI=1 S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3/i14D2 |
| SMILES | [2 H]C([2 H])(O)CCCCCCCCCCCCC |
| IUPAC Name | 1,1-dideuteriotetradecan-1-ol |
| Molecular Weight (g/mol) | 216.4 |
| Formula Weight | 216.242 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction