missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Nicotine-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | (+/-)-Nicotine-d3 |
| CAS | 69980-24-1 |
| Molecular Formula | C10 2H3 H11 N2 |
| Quantity | 0.25 g |
| Synonym | 3-[1-(Methyl-d3)-2-pyrrolidinyl]pyridine, (±)-3-[1-(Methyl-d3)-2-pyrrolidinyl]pyridine, (±)-Nicotine-N-(methyl-d3), dl-Nicotine-N-(methyl-d3) |
| InChI Formula | InChI=1 S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10 H,3,5,7H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])N1CCCC1c2cccnc2 |
| IUPAC Name | 3-[1-(trideuteriomethyl)pyrrolidin-2-yl]pyridine |
| Molecular Weight (g/mol) | 165.25 |
| Formula Weight | 165.1345 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction