missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl-d5-tin Trichloride, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Phenyl-d5-tin Trichloride |
| Molecular Formula | C6 D5 Sn . 3 Cl |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C6H5.3 ClH.Sn/c1-2-4-6-5-3-1;;;;/h1-5 H;3*1 H;/q;;;;+3/p-3/i1D,2 D,3 D,4 D,5 D;;;; |
| SMILES | [Cl-].[Cl-].[Cl-].[2 H]c1c([2 H])c([2 H])c([Sn+3 ])c([2 H])c1[2 H] |
| IUPAC Name | (2,3,4,5,6-pentadeuteriophenyl)tin(3+);trichloride |
| Molecular Weight (g/mol) | 307.204 |
| Formula Weight | 306.879 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction