missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylbenzene-d14, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | tert-Butylbenzene-D14 |
| CAS | 209963-71-3 |
| Molecular Formula | C10 D14 |
| Quantity | 0.25 g |
| Synonym | Benzene-d5, [1,1-di(methyl-d3)ethyl-2,2,2-d3]- (9 CI, ACI), [1,1-Di(methyl-d3)ethyl-2,2,2-d3]benzene-d5 (ACI), tert-Butylbenzene-d14, tert-Butylbenzene-D14 |
| InChI Formula | InChI=1 S/C10H14/c1-10(2,3)9-7-5-4-6-8-9/h4-8 H,1-3H3/i1D3,2D3,3D3,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]benzene |
| Molecular Weight (g/mol) | 148.304 |
| Formula Weight | 148.197 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction