missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-propyl-d21 Phosphate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Phosphoric Acid Tripropyl Ester-d21 |
| CAS | 1219794-92-9 |
| Molecular Formula | C9 D21 O4 P |
| Quantity | 0.25 g |
| Synonym | Tripropyl Phosphate-d21, 1-Propan-1,1,2,2,3,3,3-d7-ol, 1,1',1''-phosphate, Tri-n-propyl Phosphate D21 |
| InChI Formula | InChI=1 S/C9H21O4P/c1-4-7-11-14(10,12-8-5-2)13-9-6-3/h4-9H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OP(=O)(OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tris(1,1,2,2,3,3,3-heptadeuteriopropyl) phosphate |
| Molecular Weight (g/mol) | 245.36 |
| Formula Weight | 245.2496 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction