missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethoxybenzene-d10, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,3-Dimethoxybenzene D10 |
| CAS | 340257-57-0 |
| Molecular Formula | C8 D10 O2 |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,3,5-d4, 4,6-di(methoxy-d3)- (9 CI), 1,3-Dimethoxybenzene D10, 1,3-Dimethoxybenzene-d10, 3-Methoxyanisole-d10, 3-(Methoxy-d3)phenyl-2,4,5,6-d4 methyl-d3 ether, Resorcinol-d4 di(methyl-d3) ether, m-Dimethoxybenzene-d10, m-Methoxyanisole-d10 |
| InChI Formula | InChI=1 S/C8H10O2/c1-9-7-4-3-5-8(6-7)10-2/h3-6 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(OC([2 H])([2 H])[2 H])c([2 H])c(OC([2 H])([2 H])[2 H])c1[2 H] |
| IUPAC Name | 1,2,3,5-tetradeuterio-4,6-bis(trideuteriomethoxy)benzene |
| Molecular Weight (g/mol) | 148.225 |
| Formula Weight | 148.131 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction