missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Benzene-d4-diamine, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,4-Phenylenediamine-d4 |
| CAS | 119516-83-5 |
| Molecular Formula | C6 2H4 H4 N2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C6H8N2/c7-5-1-2-6(8)4-3-5/h1-4 H,7-8H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(N)c([2 H])c([2 H])c1N |
| IUPAC Name | 2,3,5,6-tetradeuteriobenzene-1,4-diamine |
| Molecular Weight (g/mol) | 112.17 |
| Formula Weight | 112.0939 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction