missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopentadecane-1-13C, CDN
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromopentadecane-1-13 C |
| CAS | 2708283-81-0 |
| Molecular Formula | 13 C C14 H31 Br |
| Quantity | 0.5 g |
| Synonym | 1-Bromopentadecane-1-13 C |
| InChI Formula | InChI=1 S/C15H31Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h2-15H2,1H3/i15+1 |
| SMILES | CCCCCCCCCCCCCC[13 CH2]Br |
| IUPAC Name | 1-bromo(113 C)pentadecane |
| Molecular Weight (g/mol) | 292.3033 |
| Formula Weight | 291.1643 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction