missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorophenol-3,5,6-d3, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichlorophenol-3,5,6 D3 |
| CAS | 93951-74-7 |
| Molecular Formula | C6 2H3 H Cl2 O |
| Quantity | 0.5 g |
| Synonym | Phen-2,3,5-d3-ol, 4,6-dichloro- (9 CI), 2,4-Dichlorophenol D3, 2,4-Dichlorophenol (ring-D3), 2,4-Dichlorophenol-3,5,6-D3, 2,4-Dichloro-3,5,6-trideuteriophenol |
| InChI Formula | InChI=1 S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9 H/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c([2 H])c(Cl)c1O |
| IUPAC Name | 2,4-dichloro-3,5,6-trideuteriophenol |
| Molecular Weight (g/mol) | 166.02 |
| Formula Weight | 164.9828 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction