missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-methylpropane-d9, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | tert-Butyl Bromide-D9 |
| CAS | 42310-83-8 |
| Molecular Formula | C4 D9 Br |
| Quantity | 0.5 g |
| Synonym | Propane-1,1,1,3,3,3-d6, 2-bromo-2-(methyl-d3)- (6 CI, 9 CI), tert-Butyl-d9 bromide, tert-Butyl bromide-D9, 2-Bromo-2-methylpropane-d9, 2-Bromo-2-(methyl-d3)propane-1,1,1,3,3,3-d6 |
| InChI Formula | InChI=1 S/C4H9Br/c1-4(2,3)5/h1-3H3/i1D3,2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Br)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-bromo-1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propane |
| Molecular Weight (g/mol) | 146.0737 |
| Formula Weight | 145.0453 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction