missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3,4-Dimethoxyphenyl)acetic-alpha,alpha-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-(3,4-Dimethoxyphenyl)acetic-2,2-D2 Acid |
| CAS | 19031-58-4 |
| Molecular Formula | C10 D2 H10 O4 |
| Quantity | 0.5 g |
| Synonym | Benzeneacetic-d2 acid, 3,4-dimethoxy- (9 CI), Acetic-d2 acid, (3,4-dimethoxyphenyl)- (8 CI), α,α'-Dideuteriohomoveratric acid, 3,4-Dimethoxybenzeneacetic-d2 acid, (3,4-Dimethoxyphenyl)acetic-d2 acid, 2-(3,4-Dimethoxyphenyl)acetate-2,2-d2, 2-(3,4-Dimethoxyphenyl)acetic-2,2-D2 acid, Homoveratric acid-d2, 2-(3,4-Dimethoxyphenyl)acetic acid-d2 |
| InChI Formula | InChI=1 S/C10H12O4/c1-13-8-4-3-7(6-10(11)12)5-9(8)14-2/h3-5 H,6H2,1-2H3,(H,11,12)/i6D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)c1ccc(OC)c(OC)c1 |
| IUPAC Name | 2,2-dideuterio-2-(3,4-dimethoxyphenyl)acetic acid |
| Molecular Weight (g/mol) | 198.212 |
| Formula Weight | 198.086 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction