missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzamide-2,3,4,5,6-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Benzamide-D5 |
| CAS | 1235489-47-0 |
| Molecular Formula | C7 D5 H2 N O |
| Quantity | 0.5 g |
| Synonym | Benzamide-2,3,4,5,6-d5 (ACI), Benzamide-d5, Benzene-2,3,4,5,6-d5-carboxamide, Benzoic acid amide-d5, Benzamide-D5 |
| InChI Formula | InChI=1 S/C7H7NO/c8-7(9)6-4-2-1-3-5-6/h1-5 H,(H2,8,9)/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)N)c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,5,6-pentadeuteriobenzamide |
| Molecular Weight (g/mol) | 126.1674 |
| Formula Weight | 126.0841 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction