missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocyclopropane-2,2,3,3-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Bromocyclopropane-d4 |
| CAS | 1398065-55-8 |
| Molecular Formula | C3 D4 H Br |
| Quantity | 0.5 g |
| Synonym | Cyclopropyl bromide-d4, NSC 89692-d4 |
| InChI Formula | InChI=1 S/C3H5Br/c4-3-1-2-3/h3H,1-2H2/i1D2,2D2 |
| SMILES | [2 H]C1([2 H])C(Br)C1([2 H])[2 H] |
| IUPAC Name | 3-bromo-1,1,2,2-tetradeuteriocyclopropane |
| Molecular Weight (g/mol) | 125 |
| Formula Weight | 123.9826 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction