missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorobenzene-3,5-d2, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Chlorobenzene-3,5-D2 |
| CAS | 59164-10-2 |
| Molecular Formula | C6 D2 H3 Cl |
| Quantity | 0.5 g |
| Synonym | Benzene-1,3-d2, 5-chloro- (9 CI, ACI), 5-Chlorobenzene-1,3-d2 (ACI), Chlorobenzene-3,5-d2, Chlorobenzene-3,5-D2 |
| InChI Formula | InChI=1 S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5 H/i2D,3 D |
| SMILES | [2 H]c1cc([2 H])cc(Cl)c1 |
| IUPAC Name | 1-chloro-3,5-dideuteriobenzene |
| Molecular Weight (g/mol) | 114.5692 |
| Formula Weight | 114.0205 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction