missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentene-d8, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Cyclopentene-D8 |
| CAS | 6611-46-7 |
| Molecular Formula | C5 D8 |
| Quantity | 0.5 g |
| Synonym | Cyclopentene-d8 (7 CI, 8 CI, 9 CI, ACI), Cyclopentene-D8 |
| InChI Formula | InChI=1 S/C5H8/c1-2-4-5-3-1/h1-2 H,3-5H2/i1D,2 D,3D2,4D2,5D2 |
| SMILES | [2 H]C1=C([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1,2,3,3,4,4,5,5-octadeuteriocyclopentene |
| Molecular Weight (g/mol) | 76.1663 |
| Formula Weight | 76.1128 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction