missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl-d10-amine, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
Specifications
Specifications
| Chemical Name or Material | Diphenylamine-d10 |
| CAS | 37055-51-9 |
| Molecular Formula | C12 2H10 H N |
| Quantity | 0.5 g |
| Synonym | Diphenylamine-d10, Diphenylamine D10, Benzen-2,3,4,5,6-d5-amine, N-(phenyl-2,3,4,5,6-d5)-, Benzen-d5-amine, N-(phenyl-d5)- (9 CI), N-(Phenyl-2,3,4,5,6-d5)benzen-2,3,4,5,6-d5-amine, Di(perdeuterophenyl)amine |
| InChI Formula | InChI=1 S/C12H11N/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10,13 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Nc2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 2,3,4,5,6-pentadeuterio-N-(2,3,4,5,6-pentadeuteriophenyl)aniline |
| Molecular Weight (g/mol) | 179.28 |
| Formula Weight | 179.1519 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction