missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Serine-2,3,3-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | DL-Serine-2,3,3-d3 |
| CAS | 70094-78-9 |
| Molecular Formula | C3 2H3 H4 N O3 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C3H7NO3/c4-2(1-5)3(6)7/h2,5 H,1,4H2,(H,6,7)/i1D2,2 D |
| SMILES | [2 H]C([2 H])(O)C([2 H])(N)C(=O)O |
| IUPAC Name | 2-amino-2,3,3-trideuterio-3-hydroxypropanoic acid |
| Molecular Weight (g/mol) | 108.11 |
| Formula Weight | 108.0614 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction