missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-1-d1 Alcohol, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Ethanol-1-D |
| CAS | 1624-36-8 |
| Molecular Formula | C2 D H5 O |
| Quantity | 0.5 g |
| Synonym | Ethan-1-d-ol (9 CI), Ethyl-1-d alcohol (6 CI, 7 CI, 8 CI), Ethanol-1-d, 1-Deuterioethanol, Ethanol-1-D, Ethanol-d1, Ethyl-1-d1 Alcohol |
| InChI Formula | InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3/i2D |
| SMILES | [2 H]C(C)O |
| IUPAC Name | 1-deuterioethanol |
| Molecular Weight (g/mol) | 47.0746 |
| Formula Weight | 47.0481 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction