missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetoacetate-2,2,4,4,4-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Ethyl Acetoacetate-2,2,4,4,4-D5 |
| CAS | 55514-60-8 |
| Molecular Formula | C6 D5 H5 O3 |
| Quantity | 0.5 g |
| Synonym | Butanoic-2,2,4,4,4-d5 acid, 3-oxo-, ethyl ester (9 CI), Ethyl 3-oxobutanoate-2,2,4,4,4-d5, Ethyl acetoacetate-d5, Ethyl Acetoacetate-2,2,4,4,4-D5, 1-Ethoxybutane-1,3-dione-2,2,4,4,4-d5, Acetoacetic-2,2,4,4,4-d5 acid ethyl ester, Acetoacetic acid ethyl ester-d5 |
| InChI Formula | InChI=1 S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3/i2D3,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])C(=O)OCC |
| IUPAC Name | ethyl 2,2,4,4,4-pentadeuterio-3-oxobutanoate |
| Molecular Weight (g/mol) | 135.1726 |
| Formula Weight | 135.0944 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction