missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl-d5 Trihexadecanoate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Tripalmitin-D5 (glyceryl-D5) |
| CAS | 60763-98-6 |
| Molecular Formula | C51 D5 H93 O6 |
| Quantity | 0.5 g |
| Synonym | Hexadecanoic acid, 1,2,3-propanetriyl-1,1,2,3,3-d5 ester (9 CI), Hexadecanoic acid 1,2,3-propanetriyl-1,1,2,3,3-d5 ester, Glyceryl-d5 Trihexadecanoate, Glycerol-d5 trihexadecanoate, Glyceryl-d5 tripalmitate, Trihexadecanoin-d5, Tripalmitin-d5, Tripalmitin-D5 (glyceryl-D5) |
| InChI Formula | InChI=1 S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3/i46D2,47D2,48 D |
| SMILES | [2 H]C([2 H])(OC(=O)CCCCCCCCCCCCCCC)C([2 H])(OC(=O)CCCCCCCCCCCCCCC)C([2 H])([2 H])OC(=O)CCCCCCCCCCCCCCC |
| IUPAC Name | [1,1,2,3,3-pentadeuterio-2,3-di(hexadecanoyloxy)propyl] hexadecanoate |
| Molecular Weight (g/mol) | 812.351 |
| Formula Weight | 811.7677 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction