missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanedioic-2,2,5,5-d4 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Hexanedioic Acid-2,2,5,5-D4 |
| CAS | 19031-55-1 |
| Molecular Formula | C6 D4 H6 O4 |
| Quantity | 0.5 g |
| Synonym | Hexanedioic-2,2,5,5-d4 acid (9 CI, ACI), Adipic-2,2,5,5-d4 acid (8 CI), Hexanedioic acid-2,2,5,5-D4, Hexanedioic acid-d4, Adipic Acid-d4 |
| InChI Formula | InChI=1 S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i3D2,4D2 |
| SMILES | [2 H]C([2 H])(CCC([2 H])([2 H])C(=O)O)C(=O)O |
| IUPAC Name | 2,2,5,5-tetradeuteriohexanedioic acid |
| Molecular Weight (g/mol) | 150.1659 |
| Formula Weight | 150.083 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction