missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocinnamic-d5 Acid (phenyl-d5), CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 3-Phenylpropanoic Acid D5 (phenyl-D5) |
| CAS | 35845-62-6 |
| Molecular Formula | C9 D5 H5 O2 |
| Quantity | 0.5 g |
| Synonym | Benzene-d5-propanoic acid (9 CI), 3-(Pentadeuterophenyl)propionic acid, Pentadeuterohydrocinnamic acid, 3-Phenylpropanoic acid-d5, 3-(Phenyl-d5)propanoic acid, β-Phenylpropionic-d5 acid, β-(Phenyl-d5)propionic acid, Hydrocinnamic-d5 acid, Hydrocinnamic acid-d5, Hydrocinnamic-d5 Acid (phenyl-d5), Benzenepropanoic-d5 acid |
| InChI Formula | InChI=1 S/C9H10O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5 H,6-7H2,(H,10,11)/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CCC(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | 3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid |
| Molecular Weight (g/mol) | 155.2053 |
| Formula Weight | 155.0995 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction