missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Toluic-d7 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | m-Toluic-d7 Acid |
| CAS | 733046-94-1 |
| Molecular Formula | C8 2H7 H O2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C8H8O2/c1-6-3-2-4-7(5-6)8(9)10/h2-5 H,1H3,(H,9,10)/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)O)c([2 H])c(c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(trideuteriomethyl)benzoic acid |
| Molecular Weight (g/mol) | 143.191 |
| Formula Weight | 143.096 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction