missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (±)-2-Bromopropionate-2,3,3,3-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromopropanoic Acid Methyl Ester-2,3,3,3-D4 |
| CAS | 1398065-68-3 |
| Molecular Formula | C4 D4 H3 Br O2 |
| Quantity | 0.5 g |
| Synonym | Methyl (±)-2-Bromopropionate-2,3,3,3-d4, Methyl 2-bromopropanoate-2,3,3,3-d4, 2-Bromopropanoic-2,3,3,3-d4 acid methyl ester, 2-Bromopropanoic Acid Methyl Ester-2,3,3,3-D4, 2-Bromopropionic acid-methyl ester-2,3,3,3-d4 |
| InChI Formula | InChI=1 S/C4H7BrO2/c1-3(5)4(6)7-2/h3H,1-2H3/i1D3,3 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Br)C(=O)OC |
| IUPAC Name | methyl 2-bromo-2,3,3,3-tetradeuteriopropanoate |
| Molecular Weight (g/mol) | 171.0258 |
| Formula Weight | 169.988 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction