missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Acetate-d6, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Methyl Acetate-d6 |
| CAS | 32113-85-2 |
| Molecular Formula | C3 D6 O2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C3H6O2/c1-3(4)5-2/h1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])OC(=O)C([2 H])([2 H])[2 H] |
| IUPAC Name | trideuteriomethyl |
| Molecular Weight (g/mol) | 80.12 |
| Formula Weight | 80.0744 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction