missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Decanoate-d19, CDN
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.10g
0.50g
Specifications
Specifications
| Chemical Name or Material | Decanoic Acid Methyl Ester-D19 (decanoic-D19) |
| CAS | 1219804-67-7 |
| Molecular Formula | C11 D19 H3 O2 |
| Quantity | 0.5 g |
| Synonym | Decanoic-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d19 acid methyl ester, Decanoic-d19 acid methyl ester, Methyl Decanoate-d19, Methyl Decanoate-D19, Decanoic Acid Methyl Ester-D19 (decanoic-D19), Decanoic acid methyl ester-d19 |
| InChI Formula | InChI=1 S/C11H22O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h3-10H2,1-2H3/i1D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)OC |
| IUPAC Name | methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecadeuteriodecanoate |
| Molecular Weight (g/mol) | 205.4083 |
| Formula Weight | 205.2812 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction