missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexadecyltrimethyl-d9-ammonium Bromide, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Hexadecyltrimethylammonium-d9 Bromide |
| CAS | 95217-14-4 |
| Molecular Formula | C19 2H9 H33 N . Br |
| Quantity | 0.5 g |
| Synonym | 1-Hexadecanaminium, N,N,N-tri(methyl-d3)-, bromide (1:1), 1-Hexadecanaminium, N,N,N-tri(methyl-d3)-, bromide (9 CI), Cetrimonium-D9 Bromide (N,N,N-tri(methyl-D3)) |
| InChI Formula | InChI=1 S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1 H/q+1;/p-1/i2D3,3D3,4D3; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])[N+](CCCCCCCCCCCCCCCC)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | hexadecyl-tris(trideuteriomethyl)azanium;bromide |
| Molecular Weight (g/mol) | 373.5 |
| Formula Weight | 372.3066 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction