missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-d6-aniline, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethylaniline-d6 |
| CAS | 4019-61-8 |
| Molecular Formula | C8 D6 H5 N |
| Quantity | 0.5 g |
| Synonym | Benzenamine, N,N-di(methyl-d3)- (9 CI, ACI), Aniline, N,N-di(methyl-d3)- (7 CI, 8 CI), N,N-Di(methyl-d3)benzenamine (ACI), N,N-Bis(trideuteriomethyl)aniline, N,N-Dimethyl-d6-aniline, N,N-Dimethylaniline-d6, N,N-Dimethylaniline-d6, N,N-Dimethylaniline-D6 (N,N-dimethyl-D6), N,N-Di(methyl-d3)aniline |
| InChI Formula | InChI=1 S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])N(c1ccccc1)C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 127.217 |
| Formula Weight | 127.127 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction