missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyl-d7-amine HCl, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Propylamine-D7 (propyl-D7) Hydrochloride |
| CAS | 344298-88-0 |
| Molecular Formula | C3 D7 H2 N . Cl H |
| Quantity | 0.5 g |
| Synonym | 1-Propan-1,1,2,2,3,3,3-d7-amine, hydrochloride (9 CI), Propyl-1,1,2,2,3,3,3-d7-amine hydrochloride, n-Propyl-d7-amine hydrochloride, 1-Propylamine-d7 hydrochloride, Propylamine-D7 (propyl-D7) hydrochloride |
| InChI Formula | InChI=1 S/C3H9N.ClH/c1-2-3-4;/h2-4H2,1H3;1 H/i1D3,2D2,3D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3,3-heptadeuteriopropan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 102.614 |
| Formula Weight | 102.094 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction