missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Cresol-d3 (methyl-d3), CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 2-Methylphenol-D3 (methyl-D3) |
| CAS | 70837-27-3 |
| Molecular Formula | C7H5D3O |
| Quantity | 0.5 g |
| Synonym | Phenol, 2-(methyl-d3)- (9 CI, ACI), 2-(Methyl-d3)phenol (ACI), o-Cresol-d3 (methyl-d3), 2-Methylphenol-D3 (methyl-D3) |
| InChI Formula | InChI=1 S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8 H,1H3/i1D3 |
| SMILES | OC1=CC=CC=C1C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-(methyl-d3)phenol |
| Molecular Weight (g/mol) | 111.16 |
| Formula Weight | 111.08 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction