missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Toluic Acid-13C1 (carboxyl-13C), CDN
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methylbenzoic Acid 13 C (carboxy-13 C) |
| CAS | 70838-82-3 |
| Molecular Formula | 13 C C7 H8 O2 |
| Quantity | 0.5 g |
| Synonym | Benzoic-carboxy-13 C acid, 2-methyl- (9 CI), 2-Methylbenzoic-carboxy-13 C acid, o-Toluic-13 C acid, 2-Methylbenzoic acid-13 C, 2-Methylbenzoic acid 13 C (carboxy-13 C) |
| InChI Formula | InChI=1 S/C8H8O2/c1-6-4-2-3-5-7(6)8(9)10/h2-5 H,1H3,(H,9,10)/i8+1 |
| SMILES | Cc1ccccc1[13 C](=O)O |
| IUPAC Name | 2-methyl(^{13}C)benzoic acid |
| Molecular Weight (g/mol) | 137.141 |
| Formula Weight | 137.056 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction