missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrrole-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | Pyrrole-D5 |
| CAS | 18430-85-8 |
| Molecular Formula | C4 D5 N |
| Quantity | 0.5 g |
| Synonym | 1 H-Pyrrole-1,2,3,4,5-d5 (9 CI, ACI), Pyrrole-d5 (6 CI, 8 CI), 1,2,3,4,5-Pentadeuteriopyrrole, Perdeuteriopyrrole, Pyrrole-D5 |
| InChI Formula | InChI=1 S/C4H5N/c1-2-4-5-3-1/h1-5 H/i1D,2 D,3 D,4 D/hD |
| SMILES | [2 H]c1c([2 H])c([2 H])n([2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuteriopyrrole |
| Molecular Weight (g/mol) | 72.12 |
| Formula Weight | 72.074 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction