missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Propionate-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Sodium Propionate-D5 |
| CAS | 202529-18-8 |
| Molecular Formula | C3D5NaO2 |
| Quantity | 0.5 g |
| Synonym | Propanoic-d5 acid, sodium salt (9 CI), Sodium propionate-D5, Sodium propionate-d5, Sodium propanoate-d5, Propionic acid-d5 sodium salt |
| InChI Formula | InChI=1 S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1/i1D3,2D2; |
| SMILES | O=C([O-])C([2 H])([2 H])C([2 H])([2 H])[2 H].[Na+] |
| IUPAC Name | sodium propanoate-d5 |
| Molecular Weight (g/mol) | 101.09 |
| Formula Weight | 101.05 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction