missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran-2,2,5,5-d4, CDN
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | Tetrahydrofuran D4 (2,2,5,5-D4) |
| CAS | 20665-63-8 |
| Molecular Formula | C4 D4 H4 O |
| Quantity | 0.5 g |
| Synonym | Tetrahydrofuran D4 (2,2,5,5-D4), Furan-2,5-d2, tetrahydro-2,5-d2- (6 CI, 8 CI, 9 CI, ACI), Tetrahydro-2,5-d2furan-2,5-d2 (ACI), Tetrahydrofuran-2,2,5,5-d4, Tetrahydrofuran-2,2,5,5-D4, Tetrahydrofuran-d4, THF-2,2,5,5-d4 |
| InChI Formula | InChI=1 S/C4H8O/c1-2-4-5-3-1/h1-4H2/i3D2,4D2 |
| SMILES | [2 H]C1([2 H])CCC([2 H])([2 H])O1 |
| IUPAC Name | 2,2,5,5-tetradeuteriooxolane |
| Molecular Weight (g/mol) | 76.13 |
| Formula Weight | 76.083 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction