missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethyl-d12-ammonium Chloride, CDN
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | Tetramethylammonium-D12 Chloride |
| CAS | 23789-03-9 |
| Molecular Formula | C4 D12 N . Cl |
| Quantity | 0.5 g |
| Synonym | Methan-d3-aminium, N,N,N-tri(methyl-d3)-, chloride (9 CI), Ammonium, tetra(methyl-d3)-, chloride (8 CI), Tetramethylammonium chloride-d12, Tetramethylammonium-d12 chloride, Tetramethylammonium-D12 chloride |
| InChI Formula | InChI=1 S/C4H12N.ClH/c1-5(2,3)4;/h1-4H3;1 H/q+1;/p-1/i1D3,2D3,3D3,4D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tetrakis(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 121.672 |
| Formula Weight | 121.141 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction