missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymine-alpha,alpha,alpha,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Thymine-D4 (6-D,methyl-D3) |
| CAS | 200496-79-3 |
| Molecular Formula | C5 D4 H2 N2 O2 |
| Quantity | 0.5 g |
| Synonym | 2,4(1 H,3 H)-Pyrimidinedione-6-d, 5-(methyl-d3)- (9 CI, ACI), 5-(Methyl-d3)-2,4(1 H,3 H)-pyrimidinedione-6-d (ACI), Thymine-d4, Thymine-D4 (6-D,methyl-D3), Thymine-α,α,α,6-d4 |
| InChI Formula | InChI=1 S/C5H6N2O2/c1-3-2-6-5(9)7-4(3)8/h2H,1H3,(H2,6,7,8,9)/i1D3,2 D |
| SMILES | [2 H]C1=C(C(=O)NC(=O)N1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 6-deuterio-5-(trideuteriomethyl)-1 H-pyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 130.138 |
| Formula Weight | 130.068 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction