missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-butyl-d27 Phosphate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | Tributyl phosphate-d27 |
| CAS | 61196-26-7 |
| Molecular Formula | C12 D27 O4 P |
| Quantity | 0.5 g |
| Synonym | 1-Butan-1,1,2,2,3,3,4,4,4-d9-ol, phosphate (3:1) (9 CI), Tributyl Phosphate D27 |
| InChI Formula | InChI=1 S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OP(=O)(OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tris(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl) phosphate |
| Molecular Weight (g/mol) | 293.48 |
| Formula Weight | 293.334 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction