missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroethylene-d, CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 g
1 g
missing translation for 'unitSize'
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Trichloroethylene-d |
| CAS | 13291-68-4 |
| Molecular Formula | C2 D Cl3 |
| Quantity | 0.5 g |
| Synonym | Ethene-d, trichloro- (9 CI), Ethylene-d, trichloro- (6 CI, 7 CI, 8 CI), 1,1,2-Trichloro-2-deuterioethene, Deuterotrichloroethylene |
| InChI Formula | InChI=1 S/C2HCl3/c3-1-2(4)5/h1H/i1D |
| SMILES | [2 H]C(=C(Cl)Cl)Cl |
| IUPAC Name | 1,1,2-trichloro-2-deuterioethene |
| Molecular Weight (g/mol) | 132.3945 |
| Formula Weight | 130.9207 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction