missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Canine IgG Fc Secondary Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72773C
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
| IgG Fc | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Dog IgG F(c) fragment | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Canine | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| DyLight 650 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction