missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane, 97%, Thermo Scientific™

Product Code. 10561531 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10561531 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10561531 Supplier Thermo Scientific Acros Supplier No. 312440050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-57-4
Molecular Formula C8H4F13I
Molecular Weight (g/mol) 474.004
MDL Number MFCD00039410
InChI Key NVVZEKTVIXIUKW-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodo, 1-iodo-1h,1h,2h,2h-perfluorooctane, 1,1,2,2-tetrahydroperfluorooctyliodide, 1,1,2,2-tetrahydroperfluorooctyl iodide, 2-perfluorohexyl ethyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-iodooctane, 1h,1h,2h,2h-tridecafluoro-n-octyl iodide, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl iodide, acmc-1ccsn
PubChem CID 74888
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-8-iodooctane
SMILES C(CI)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.9340g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ICH2CH2(CF2)5CF3
Refractive Index 1.3590 to 1.3610 (20°C, 589nm)
Quantity 5 g
Formula Weight 474
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodooctane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.