missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,11,13,13,13-Octachlorotridecane, TRC

Product Code. 30007197
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007197 5 mg 5mg
30007276 50 mg 50mg
30008322 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007197 Supplier Toronto Research Chemicals Supplier No. TRCO2336135MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865306-24-7
Molecular Formula C13 H20 Cl8
Molecular Weight (g/mol) 459.92
Synonym Tridecane, 1,1,1,3,11,13,13,13-octachloro- (9CI, ACI), 1,1,1,3,11,13,13,13-Octachlorotridecane (ACI)
IUPAC Name 1,1,1,3,11,13,13,13-octachlorotridecane
SMILES ClC(CCCCCCCC(Cl)CC(Cl)(Cl)Cl)CC(Cl)(Cl)Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H20Cl8/c14-10(8-12(16,17)18)6-4-2-1-3-5-7-11(15)9-13(19,20)21/h10-11H,1-9H2
Formula Weight 455.9073
Chemical Name or Material 1,1,1,3,11,13,13,13-Octachlorotridecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.