missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,11,13,13,13-Octachlorotridecane, TRC

Product Code. 30008322
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008322 25 mg 25mg
30007197 5 mg 5mg
30007276 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008322 Supplier Toronto Research Chemicals Supplier No. TRCO23361325MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 865306-24-7
Molecular Formula C13 H20 Cl8
Molecular Weight (g/mol) 459.92
Synonym Tridecane, 1,1,1,3,11,13,13,13-octachloro- (9CI, ACI), 1,1,1,3,11,13,13,13-Octachlorotridecane (ACI)
IUPAC Name 1,1,1,3,11,13,13,13-octachlorotridecane
SMILES ClC(CCCCCCCC(Cl)CC(Cl)(Cl)Cl)CC(Cl)(Cl)Cl

Especificaciones

Quantity 25 mg
InChI Formula InChI=1S/C13H20Cl8/c14-10(8-12(16,17)18)6-4-2-1-3-5-7-11(15)9-13(19,20)21/h10-11H,1-9H2
Formula Weight 455.9073
Chemical Name or Material 1,1,1,3,11,13,13,13-Octachlorotridecane
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.