missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol, 98+%

Product Code. 11459307
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459307 1 g 1g
11469307 5 g 5g
11479307 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459307 Supplier Thermo Scientific Alfa Aesar Supplier No. B21366.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 718-64-9 | C9H6F6O | 244.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 718-64-9
Molecular Formula C9H6F6O
Molecular Weight (g/mol) 244.136
MDL Number MFCD00040983
InChI Key IZPIZCAYJQCTNG-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-phenyl-2-propanol, hexafluoro-2-hydroxyisopropyl benzene, bis trifluoromethyl phenylcarbinol, 2,2,2,2',2',2'-hexafluorocumyl alcohol, alpha,alpha-bis trifluoromethyl benzyl alcohol, hexafluoro-2-phenylisopropanol, benzenemethanol, .alpha.,.alpha.-bis trifluoromethyl, hexafluoro-2-phenyl isopropanol, hexafluoro-2-phenyl-2-propanol, benzenemethanol, alpha,alpha-bis trifluoromethyl
PubChem CID 69748
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-ol
SMILES C1=CC=C(C=C1)C(C(F)(F)F)(C(F)(F)F)O

Specifications

Density 1.45
Boiling Point 159°C to 160°C
Flash Point 60°C (140°F)
Refractive Index 1.415
Quantity 1 g
UN Number UN1987
Beilstein 2053547
Formula Weight 244.14
Percent Purity ≥98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.