missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis

Product Code. 12655007 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12655007 10 mL Glass bottle 10mL
12665007 50 mL Glass bottle 50mL
15189303 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Product Code. 12655007 Supplier Thermo Scientific Acros Supplier No. 445820100
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

CAS: 920-66-1 | C3H2F6O | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-66-1
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.04
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifications

Melting Point -4.0°C
Color White to Yellow
Density 1.5960g/mL
Boiling Point 59.0°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CF3)2CHOH
Refractive Index 1.2740 to 1.2770
Quantity 10 mL
Residue after Evaporation 0.001 % max.
Specific Gravity 1.596
Solubility Information Solubility in water: 1000g/L (25°C)
Viscosity 1.65 mPa.s (20°C)
Formula Weight 168.04
Percent Purity 99%
Grade Analysis
Physical Form Solid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,1,1,3,3,3-Hexafluoro-2-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.