missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroacetone hydrate, 98%, Thermo Scientific™

Product Code. 15510534 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15510534

Brand: Thermo Scientific Acros 312120050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10057-27-9
Molecular Formula C3F6O
Molecular Weight (g/mol) 166.02
MDL Number MFCD00149063
InChI Key VBZWSGALLODQNC-UHFFFAOYSA-N
Synonym hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
PubChem CID 36719
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
SMILES FC(F)(F)C(=O)C(F)(F)F

Specifications

Melting Point 18.0°C to 21.0°C
Density 1.5800g/mL
Boiling Point 105.0°C to 108.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 1.58
Solubility Information Solubility in water: soluble.
Formula Weight 166.02
Percent Purity 98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroacetone hydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexafluoroacetone hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.